Learn More
Ace Glass Impinger, Greenburg-Smith, Modified
A high-velocity impinger for the determination of dust concentration in air or other gases.
Supplier: Ace Glass 753632
Description
Same as 7536-10, except tube is modified with spherical joints, both vertical, for connecting in series. Bottle joint is 45/50.Specifications
Each | |
Impinger |