Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133, Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP244248AF594

Catalog No. NB007039


Add to cart

Description

Description

CD133 Polyclonal antibody specifically detects CD133 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

CD133
Alexa Fluor 594
50mM Sodium Borate with 0.05% Sodium Azide
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Rabbit
IgG
0.1 mL
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary
Immunogen has 100% homology to all isoforms of human CD133.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Affinity Purified, Conjugated
PROM1
A synthetic peptide made to an internal portion of the human CD133 protein (between residues 600-700) [UniProt O43490]
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit