Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD134 Mouse anti-Human, BB700, Clone: ACT35 (also known as Ber-ACT35), BD Horizon™

Mouse Monoclonal Antibody

$175.00 - $385.00

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen CD134
Conjugate BB700
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566560
View Documents
BD Biosciences
566560
25 Tests Each for $175.00
Add to cart
 
BDB566559
View Documents
BD Biosciences
566559
100 Tests Each for $385.00
Add to cart
 
Description

Description

CD134 Monoclonal antibody specifically detects CD134 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Monoclonal
Primary
CD134
BB700
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit