Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD135 (Flt3) Rat anti-Mouse, Biotin, Clone: A2F10, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 13135181

 View more versions of this product

Catalog No. 50-117-56


Add to cart

Description

Description

Description: The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively. Applications Reported: The A2F10 antibody has been reported for use in flow cytometric analysis. Applications Tested: The A2F10 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 µm post-manufacturing filtered.

This gene encodes a class III receptor tyrosine kinase that regulates hematopoiesis. The receptor consists of an extracellular domain composed of five immunoglobulin-like domains, one transmembrane region, and a cytoplasmic kinase domain split into two parts by a kinase-insert domain. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.
Specifications

Specifications

CD135 (Flt3)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q00342
RP11-153M24.3, CD135, FLK2, STK1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
A2F10
Biotin
Flt3
Liquid
Flt3
IgG2a, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14255
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.