Learn More
CD137 (4-1BB) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ CD137 (4-1BB) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS621B
Description
4-1BB (also known as CD137) is an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. It exists as both a monomer and a dimer on the surface of activated T cells. 4-1BB is structurally related to the members of NGFR/TNFR superfamily which are characterized by the presence of three-six patterns of a cysteine-rich motif in their extracellular domains. Other members of this family include low affinity NGFR, two receptors for TNF (TNFR-I & TNFR-II), CD30, CD40, OX40, Fas, and CD27. These molecules are involved in cell growth, survival, and death processes. The cytoplasmic domain of 4-1BB include two runs of acidic amino acids, a potential p56lck binding site, five consecutive glycines at the C-terminus, and four potential phosphorylation sites: one tyrosine, two threonine, and one serine.Host Species: Mouse
Clone: BBK-2
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Ectodomain
Immunogen: Ectodomain of human 4-1BB recombinant protein
Molecular Weight: 30kDa
Positive Control: Spleen, thymus or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD137 Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Apoptosis | |
Human | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
BBK-2 | |
Biotin | |
Ectodomain of human 4-1BB recombinant protein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.