Learn More
CD137 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Supplier: Pepro Tech Inc 500P167G1MG

Description
AA Sequence of recombinant protein: MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human 4-1BB Receptor. Anti-Human 4-1BB Receptor-specific antibody was purified by affinity chromatography employing an immobilized Human 4-1BB Receptor matrix. Sandwich ELISA: To detect Human 4-1BB Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human 4-1BB Receptor (500-P167GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human 4-1BB Receptor. Western Blot: To detect Human 4-1BB Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human 4-1BB Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
CD137 Polyclonal antibody specifically detects CD137 in Human samples. It is validated for ELISA, Western BlotSpecifications
CD137 | |
Polyclonal | |
PBS with no preservative | |
Q07011 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3604 | |
-20°C |
ELISA, Western Blot | |
Unconjugated | |
TNFRSF9 | |
4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9 | |
E.coli-derived Recombinant Human 4-1BB Receptor | |
1 mg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.