Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138 Mouse anti-Human, APC-R700, Clone: MI15  , BD Horizon™

Mouse Monoclonal Antibody

$144.60 - $347.50

Specifications

Antigen CD138
Isotype Mouse BALB/c IgG1 κ
Immunogen Human U266 and XG-1 Myeloma Cell Lines
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Conjugate APC-R700
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566051
View Documents
BD Biosciences
566051
25 Tests
Each for $144.60
Add to cart
 
BDB566050
View Documents
BD Biosciences
566050
100 Tests
Each for $347.50
Add to cart
 
Description

Description

CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.

CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD138
Human U266 and XG-1 Myeloma Cell Lines
APC-R700
Cell Biology
Mouse
Purified
Syndecan; SDC; Syndecan 1; SDC1; SYND1
Vol. Per test: 5μL
Mouse BALB/c IgG1 κ
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
MI15  
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit