Learn More
CD138 Mouse anti-Human, BV421, Clone: MI15, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565943
Catalog No. BDB565943
Add to cart
Description
Description
CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD138
MI15
Vol. Per test: 5μL
Purified
Mouse
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Syndecan; SDC; Syndecan 1; SDC1; SYND1
Human U266 and XG-1 Myeloma Cell Lines
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.