Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138 Mouse anti-Human, PE, Clone: 1D4, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MHCD13804

Catalog No. 50-112-1340


Add to cart

Description

Description

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.

CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
Specifications

Specifications

CD138
Monoclonal
PE
SDC1
Liquid
SDC1
Human CD138
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
1D4
PBS with sucrose and 0.1% sodium azide
P18827
SDC1, SYND1, SDC, CD antigen 138
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
6382
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit