Learn More
CD14 Mouse anti-Human, Alexa Fluor™ 594, Clone: RPA-M1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227215AF594
Catalog No. NP22721A94
Add to cart
Description
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
CD14
RPA-M1
Flow Cytometry, Immunocytochemistry/Immunofluorescence
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD14
Human PHA-stimulated PBMC were used as immunogen.
0.1 mL
Primary
929