Learn More
CD14 Mouse anti-Human, Clone: 4AW4, Abnova™
Mouse monoclonal antibody raised against human CD14.
Manufacturer:  Abnova Corporation MAB12389
Catalog No. 89-933-744
Add to cart
Description
Description
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq]
Specifications
Specifications
CD14
4AW4
Mouse monoclonal antibody raised against human CD14.
In PBS, pH 7.4 (0.09% sodium azide)
CD14
CD14
Protein A/G affinity chromatography
RUO
Primary
929
Flow Cytometry, Immunofluorescence
Unconjugated
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG1
100 uL
Store at 4°C. Do not freeze.
Monoclonal
Human