Learn More
CD14 Mouse anti-Human, Porcine, Bovine, Canine, Feline, Goat, Several Species, Alexa Fluor™ 594, Clone: Tuk4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP222153AF594
Catalog No. NP22213A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD14 Monoclonal antibody specifically detects CD14 in Human,Porcine,Bovine,Canine,Feline,Goat,Llama,Mustelid,Primate,Rabbit,Sheep samples. It is validated for Western Blot,Flow Cytometry,ELISA.
Specifications
Specifications
CD14
Tuk4
Western Blot, Flow Cytometry, ELISA
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
This antinody recognises the human CD14 cell surface antigen, expressed strongly by most peripheral blood monocytes and weakly on granulocytes. Clone Tuk4 has been shown to block SDF-induced chemotaxis of U937 cells in a dose-dependent manner, see Yang, H. et al. for details.
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD14
Made to Human CD14
0.1 mL
Primary
929
Bovine, Canine, Feline, Goat, Human, Llama, Mustelid, Porcine, Primate, Rabbit, Sheep