Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Rabbit anti-Human, Alexa Fluor 647, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289260AF647

Catalog No. NB066045

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD14
2
Flow Cytometry
Purified
Rabbit
IgG
0.1 mL
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
Primary
929
Flow Cytometry
Alexa Fluor 647
50mM Sodium Borate
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD14 (NP_000582.1; Met1-Met344).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit