Learn More
CD14 Rat anti-Mouse, BUV737, Clone: rmC5-3, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565309
Catalog No. BDB565309
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD14 Monoclonal antibody specifically detects CD14 in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
CD14
rmC5-3
Brilliant Ultraviolet 737
Purified
Rat
LOU/C
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BUV737 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Cd14; CD14 antigen; Myeloid cell-specific leucine-rich glycoprotein
Recombinant Mouse CD14
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.