Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Rat anti-Mouse, FITC, Clone: Sa2-8, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 11014181

 View more versions of this product

Catalog No. 50-945-1


Add to cart

Description

Description

Description: The Sa2-8 monoclonal antibody reacts with mouse CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is a receptor for the complexes of LPS and LBP (LPS-Binding Protein) and is shown to associate with Toll-Like Receptor 4 (TLR4) and participate in the signaling and cellular response to bacterial LPS. In mouse, CD14 is expressed on the surface of macrophages and under certain conditions is also found in the serum in a secreted form. Sa2-8 has weak antagonistic activity (in NF-kappaB activation or TNF alpha production with LPS stimulation). Applications Reported: The Sa2-8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Sa2-8 antibody has been tested by flow cytometric analysis of thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifications

Specifications

CD14
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P10810
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
Sa2-8
FITC
Cd14
Liquid
Cd14
IgG2a, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12475
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.