Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD140b (PDGFRB) Mouse anti-Human, Clone: PR7212, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA528128

Catalog No. PIMA528128


Add to cart

Description

Description

This antibody recognizes the extracellular domain of the human PDGF receptor beta subunit.

PDGFRB is involved in Premature aging syndrome, Penttinen type (PENTT). A syndrome characterized by a prematurely aged appearance with lipoatrophy, epidermal and dermal atrophy along with hypertrophic lesions that resemble scars, thin hair, proptosis, underdeveloped cheekbones, and marked acro-osteolysis. The disease is caused by mutations affecting the gene represented in this entry.
Specifications

Specifications

CD140b (PDGFRB)
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P09619
PDGFRB
Human skin fibroblast cell membrane extracts
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
PR7212
Unconjugated
PDGFRB
Liquid
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5159
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit