Learn More
CD142 Mouse anti-Human, Brilliant Violet 786, Clone: HTF-1, BD Optibuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 744008
Description
The HTF-1 monoclonal antibody specifically binds to CD142. CD142 is a 45-47 kDa, single chain, type I transmembrane glycoprotein also known as Tissue Factor (TF). CD142 has been referred in the literature as coagulation Factor III or thromboplastin and it is expressed on activated endothelial cells and lipopolysaccharide (LPS)-stimulated monocytes/macrophages. TF associates with factor VIIa to form a complex and acts as an enzyme that initiates the blood coagulation cascade. It is known as the major initiator of clotting in normal hemostasis. CD142 can be induced by various inflammatory mediators in monocytes and vascular endothelial cells. This antibody is useful in inflammation, thrombosis and hemostasis research.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.Specifications
CD142 | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Tissue factor, TF, TFA, Coagulation factor III, F3, Thromboplastin | |
Mouse IgG1, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
HTF-1 | |
Brilliant Violet 786 | |
Affinity Purified | |
Murine | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV786 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.