Learn More
CD146 Mouse anti-Human, Brilliant Violet 650, Clone: P1H12, BD Optibuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 743302
Description
The P1H12 monoclonal antibody specifically binds to CD146. CD146 is a 118 kDa transmembrane glycoprotein also known as MCAM, MUC18, or Mel-CAM. CD146 is a member of the immunoglobulin superfamily strongly expressed by blood vessel endothelium and smooth muscle. CD146 is also expressed by angioblasts, mesenchymal stem cells, melanoma cells, intermediate trophoblasts and a subpopulation of activated T cells. The P1H12 monoclonal antibody has been reported to block endothelial cell adhesion that is observed very early in embryogenesis. It can be useful in the study of embryologic vasculogenesis. This antibody is suitable for immunohistochemical staining of acetone-fixed frozen tissue sections, immunoprecipitation and ELISA.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.Specifications
CD146 | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
MCAM, MELCAM, MUC18, Gicerin, Melanoma cell adhesion molecule | |
Human Umbilical Vein Cells | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV650 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Human |
P1H12 | |
Brilliant Violet 650 | |
Affinity Purified | |
Murine | |
Mouse IgG1, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.