Learn More
CD147 Mouse anti-Human, FITC, Clone: MEM-M6/1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15740
Catalog No. 50-113-7740
Add to cart
Description
Description
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and maligt tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
Specifications
Specifications
CD147
Monoclonal
FITC
BSG
Liquid
BSG
Human CD147
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-M6/1
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P35613
5F7, CD147, EMMPRIN, M6, OK, TCSF, CD147 antigen, OK blood group antigen, basigin, collagenase stimulatory factor, extracellular matrix metalloproteinase inducer, leukocyte activation antigen M6, tumor cell-derived collagenase stimulatory factor
Mouse
IgG1
100 tests
4° C
Primary
682