Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD15 Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Thermo Scientific™ CD15 Ab-3, Mouse Monoclonal Antibody.

$157.00 - $543.00

Specifications

Antigen CD15 Ab-3
Clone MMA; same as LeuM1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 8
Catalog Number Mfr. No. Concentration Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Format Quantity Price Quantity & Availability  
MS1259P
View Documents
Thermo Scientific™
MS1259P
200μg/mL Concentrated 1mL
Each for $540.00
Add to cart
 
MS1259P0
View Documents
Thermo Scientific™
MS1259P0
200μg/mL Concentrated 100μL
Each for $157.00
Add to cart
 
MS1259P1
View Documents
Thermo Scientific™
MS1259P1
200μg/mL Concentrated 500μL
Each for $360.00
Add to cart
 
MS1259P1ABX
View Documents
Thermo Scientific™
MS1259P1ABX
1mg/mL Concentrated 100μL
Each for $354.50
Add to cart
 
MS1259PABX
View Documents
Thermo Scientific™
MS1259PABX
1mg/mL Concentrated 200μL
Each for $440.00
Add to cart
 
MS1259R7
View Documents
Thermo Scientific™
MS1259R7
---- Prediluted 7mL
Each for $412.00
Add to cart
 
MS1259RQ
View Documents
Thermo Scientific™
MS1259RQ
---- Ready to Use 12mL
Each for $543.00
Add to cart
 
MS1259R1
View Documents
Thermo Scientific™
MS1259R1
---- Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Thermo Scientific™ CD15 Ab-3, Mouse Monoclonal Antibody.

3-fucosyl-N-acetyllactosamine (3-FAL) or CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: MMA; same as LeuM1

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: U937 histiocytic cell line

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Cellular Localization: Membrane and granular paranuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Specifications

CD15 Ab-3
Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
MMA; same as LeuM1
Monoclonal
Murine
Oncogenesis
U937 histiocytic cell line
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit