Learn More
CD15/Lewis X Mouse anti-Human, Alexa Fluor 405, Clone: 6B11B4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237339AF405
Catalog No. NBP237339N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD15/Lewis X
6B11B4
Western Blot, ELISA
Purified
FUT4
Purified recombinant fragment of human FUT4 (AA: 199-302) expressed in E. Coli.
Protein A or G purified
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers
Primary
2526
Western Blot, ELISA
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human