Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD15/Lewis X Mouse anti-Human, Clone: ARE, Novus Biologicals™
Mouse Monoclonal Antibody
$155.40 - $397.00
Specifications
Antigen | CD15/Lewis X |
---|---|
Clone | ARE |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD15/Lewis X Monoclonal antibody specifically detects CD15/Lewis X in Human samples. It is validated for Flow CytometrySpecifications
CD15/Lewis X | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
2526 | |
IgM | |
Protein A or G purified |
ARE | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
CD 15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
Human CD15/Lewis X (Uniprot: P22083) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title