Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15/LewisX Mouse anti-Human, Janelia Fluor 646, Clone: HI98, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP139914JF646

Catalog No. NBP13914646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD15/LewisXMonoclonalantibody specifically detects CD15/LewisX in Human samples.
Specifications

Specifications

CD15/LewisX
HI98
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Affinity Purified
FUT4
Human PBMCs were used as immunogen.
Immunogen affinity purified
Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers
Primary
2526
Human, Primate
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Mouse
IgM
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone HI98 has been described to specifically react with 3-fucosyl-N-acetyllactosamine (3-FAL), also called X-hapten, SSEA-1, CD15, or Lewis X.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit