Learn More
CD15 Monoclonal Antibody (HI98), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$227.00
Specifications
Antigen | CD15 |
---|---|
Clone | HI98 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-114-46
|
Invitrogen
13015982 |
100 μg |
Each of 1 for $227.00
|
|
|||||
Description
Description: The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative. Applications Reported: The HI98 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen and FFPE tissue sections (IHC-F, IHC-P). Applications Tested: The HI98 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.Specifications
CD15 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
stage-specific embryonic antigen 1; CD15; SSEA-1; SSEA1; ELFT; FCT3A; FUC-TIV; FUTIV; LeX | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
HI98 | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Biotin | |
Mouse | |
Human | |
IgM κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.