Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15 Mouse anti-Human , BV421, Clone: HI98 (also known as HIM1), BD Biosciences™

Mouse Monoclonal Antibody

$205.00 - $448.50

Specifications

Antigen CD15
Clone HI98 (also known as HIM1)
Applications Flow Cytometry
Conjugate Brilliant Violet 421
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB567009
View Documents
BD Biosciences
567009
25 Tests
Each for $205.00
Add to cart
 
BDB567008
View Documents
BD Biosciences
567008
100 Tests
Each for $448.50
Add to cart
 
Description

Description

CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD15
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Mononuclear Cells
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
HI98 (also known as HIM1)
Brilliant Violet 421
Mouse
Human
Lewis X; Le-X; X-Hapten; SSEA-1; 3-FAL
Mouse IgM, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit