Learn More
CD151 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0160-6F40, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212144AF594
Catalog No. NP212144A94
Add to cart
Description
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD151
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
CD151
Human CD151
Primary
Human
IgG1
Flow Cytometry
MM0160-6F40
Flow Cytometry
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151
Mouse
0.1 mL
977
Store at 4°C in the dark.