Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Monoclonal Antibody (A6G11), Huabio
Mouse Monoclonal Antibody
Supplier: HUABIO RESEARCH INC HA600059


Description
CD151 Monoclonal antibody specifically detects CD151 in Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
CD151 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O35566, P48509 | |
Mouse | |
Protein A | |
RUO | |
12476, 977 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
A6G11 | |
Unconjugated | |
CD151 | |
CD 151; CD151; CD151 antigen; CD151 antigen (Raph blood group); CD151 molecule; CD151 molecule (Raph blood group); GP27; hemidesmosomal tetraspanin CD151; membrane glycoprotein SFA-1; MER2; Peta3; PETA-3; platelet endothelial tetraspan antigen-3; platelet surface glycoprotein gp27; platelet-endothelial cell tetraspan antigen 3; platelet-endothelial tetraspan antigen 3; RAPH; SFA1; SFA-1; Tetraspanin-24; TSPAN24; tspan-24 | |
Recombinant protein within human CD151 aa 104-253. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction