Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0160-6F40, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212144AF532
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Flow Cytometry.Specifications
CD151 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD151 | |
Human recombinant CD151 | |
Primary | |
Human | |
IgG2 |
Flow Cytometry | |
MM0160-6F40 | |
Flow Cytometry | |
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
Mouse | |
0.1 mL | |
977 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title