Learn More
CD151 Rat anti-Mouse, Alexa Fluor 488, Clone: 455807, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB4609G100UG
Catalog No. FAB4609G100
Add to cart
This item is not returnable. View return policy
Description
Description
CD151 Monoclonal specifically detects CD151 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD151
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
BAA22447
CD151
HEK293 human embryonic kidney cell line transfected with mouse CD151 Met1-Tyr253 Accession # BAA22447
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse CD151. Stains mouse CD151 transfectants but not irrelevant transfectants.
Flow Cytometry
455807
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD151, CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, SFA-1, tetraspanin-24, tspan-24, TSPAN24, TSPAN24hemidesmosomal tetraspanin CD151
Rat
IgG2a
RUO
Primary
977
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)