Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD152 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB700, Clone: BNI3, BD Biosciences™

Mouse Monoclonal Antibody

$210.50 - $437.50

Specifications

Antigen CD152
Clone BNI3
Applications Flow Cytometry
Conjugate BB700
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566902
View Documents
BD Biosciences
566902
25 Tests
Each for $210.50
Add to cart
 
BDB566901
View Documents
BD Biosciences
566901
100 Tests
Each for $437.50
Add to cart
 
Description

Description

CD152 Monoclonal antibody specifically detects CD152 in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow Cytometry
Specifications

Specifications

CD152
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CTLA4 Recombinant Protein
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
BNI3
BB700
Mouse
Human, Rhesus Monkey, Cynomolgus, Baboon
CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4
Mouse BALB/c IgG2a, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit