Learn More
CD153 Rat anti-Mouse, BB700, Clone: RM153, BD Optibuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 742188
Catalog No. BDB742188
Add to cart
Description
Description
The RM153 monoclonal antibody specifically recognizes a 40-kDa type-II membrane glycoprotein, CD30 ligand (CD30L or CD153), which belongs to the NGF/TNF superfamily. In the presence of cytokines, CD153 stimulates B-cell proliferation, antigen-specific antibody production, and polyclonal immunoglobulin secretion. In addition, it costimulates the proliferation of activated T cells. This molecule is expressed on activated T cells, predominantly CD4+, on activated EL4 cells, and on activated Th1 and Th2 cells. The RM153 mAb inhibits the binding of CD153 to CD30.
The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
Specifications
Specifications
CD153
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CD30 Ligand, Cd30l, CD30LG, Tnfsf8
CHO cells transfected with mouse CD153 gene (Tnfsf8)
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BB700 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
RM153
BB700
Affinity Purified
Rat
Rat SD, also known as Sprague-Dawley (outbred) IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal