Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD154 (CD40 Ligand) Chimeric Recombinant Rabbit Monoclonal Antibody (MR1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548163
Description
Specificity: This antibody is specific for murine CD40 ligand (CD40L), a 39 kDa transmembrane glycoprotein.
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
Specifications
CD154 (CD40 Ligand) Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P27548 | |
CD40LG | |
This antibody was raised by immunising Armenian hamsters with murine activated Th1 (D1.6) plasma membrane as described by Noelle et al (1992). | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG κ |
Flow Cytometry, Functional Assay, Inhibition Assays | |
MR1 | |
Unconjugated | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Rabbit | |
Protein A | |
RUO | |
21947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction