Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Supplier: Invitrogen HMCD15401
Description
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD154 (CD40 Ligand) | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P27548 | |
CD40LG | |
Mouse CD154. from Activated mouse Th1 clone D1.6 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG3 |
Flow Cytometry | |
MR1 | |
FITC | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Armenian Hamster | |
purified | |
RUO | |
21947 | |
4° C, store in dark | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction