Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Rabbit anti-Rat, Biotin, Clone: 26, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290836B
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Rat samples. It is validated for ELISA.Specifications
CD155/PVR | |
Monoclonal | |
Biotin | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD155/PVR (NP_058772.2; Met1-Gly352). | |
0.1 mL | |
Virology Bacteria and Parasites | |
5817 | |
Rat | |
Purified |
ELISA | |
26 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human PVRL1 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title