Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Rat anti-Mouse, Alexa Fluor 647, Clone: 690912, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB6909R100UG
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Mouse samples. It is validated for Flow Cytometry.Specifications
CD155/PVR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_081790 | |
PVR | |
Mouse myeloma cell line NS0-derived recombinant mouse CD155/PVR Asp29-Leu348 Accession # NP_081790 | |
RUO | |
5817 | |
Mouse | |
IgG2a |
Flow Cytometry | |
690912 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CD155/PVR in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)