Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD156c Mouse anti-Human, PE, Clone: 11G2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 566235

Catalog No. BDB566235


Add to cart

Description

Description

The 11G2 monoclonal antibody specifically binds to CD156c which is also known as Disintegrin and metalloproteinase domain-containing protein 10 (ADAM10). CD156c is a ñ70 kDa type I transmembrane glycoprotein that belongs to the ADAM family. It is widely expressed on hematopoietic and non-hematopoietic cells. CD156c is expressed on the plasma membrane, as well as by membranes within intracellular compartments and exosomes. CD156c serves as a broadly-reactive endopeptidase that cleaves membrane-bound proteins in a process known as ectodomain shedding or release. It can cleave transmembrane molecules into soluble forms and thereby regulate the functions of a variety of different receptors and ligands that are involved in cellular signaling and adhesion. These include membrane TNF, CX3CL1, CXCL16, Ephrin-A2, Notch and Delta molecules, cadherins, CD23, CD44, IL-6 Receptor/CD126, CD171, and amyloid precursor protein. CD156c expression may be constitutive or induced, such as in inflamed tissues, including arthritic joints and in the nervous system. Dysregulated CD156c expression may be associated with certain cancers and Alzheimer’s disease.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
Specifications

Specifications

CD156c
PE
Affinity Purified
Murine
Mouse BALB/c IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
11G2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
ADAM 10, AD10, ADA10, MADM, KUZ, Kuzbanian protein homolog, HsT18717, RAK
Human Jurkat Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit