Learn More
CD157 Mouse anti-Human, BV421, Clone: SY/11B5, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564869
Catalog No. BDB564869
Add to cart
Description
Description
CD157 Monoclonal antibody specifically detects CD157 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD157
SY/11B5
Vol. Per test: 5μL
Purified
Mouse
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
BST1; Bone marrow stromal antigen 1; ADP-ribosyl cyclase 2; cADPr hydrolase
Human CD157 Transfected Cell Line
50 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.