Learn More
CD158b Monoclonal Antibody (DX27), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 605010

Description
Positive control: Tonsil Cellular location: Membrane, cytoplasm.
CD158b Monoclonal antibody specifically detects CD158b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
CD158b | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P43628 | |
Mouse | |
Protein A | |
RUO | |
3804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
DX27 | |
Unconjugated | |
KIR2DL3 | |
CD158 antigen-like family member B2; CD158b; CD158B2; CU464054.1; GL183; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 2DL3; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 3; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig-like inhibitory receptor; killer inhibitory receptor cl 2-3; KIR-023GB; KIR2DL3; KIR2DS3; KIR2DS5; KIRCL23; KIR-K7b; KIR-K7c; MGC129943; MHC class I NK cell receptor; natural killer associated transcript 2; natural killer cell inhibitory receptor KIR2DL3; natural killer cell receptor; natural killer-associated transcript 2; NKAT; NKAT2; NKAT-2; NKAT2A; NKAT2B; NK-receptor; p58; p58 natural killer cell receptor clone CL-6; p58 NK receptor CL-6; p58.2 MHC class-I specific NK receptor; p58.2 MHC class-I-specific NK receptor | |
A recombinant protein of human CD158b | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.