Learn More
CD16/CD32 Rat anti-Mouse, Alexa Fluor 405, Clone: 93, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127946AF405
Catalog No. NBP127946AF
Add to cart
Description
Description
CD16/CD32 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
CD16/CD32
93
Flow Cytometry, Immunohistochemistry-Frozen
Purified
FCGR3A
Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Protein A or G purified
RUO
Primary
2214
Mouse
Flow Cytometry, Immunohistochemistry (Frozen)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Rat
IgG2a
0.25 ml
Store at 4C in the dark.
Monoclonal
Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)