Learn More
CD16/CD32, Rat anti-Mouse, Clone: 93, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP280628
Catalog No. NB013701
Add to cart
Description
Description
CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
Specifications
CD16/CD32
Monoclonal
0.5 mg/mL
Flow Cytometry : 3 ug/10^6 cells, Immunohistochemistry, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, Block/Neutralize
CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Rat
Protein A or G purified
RUO
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Blocking Assay
93
Unconjugated
PBS
FCGR3A
Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
0.25 mg
Adaptive Immunity, Immunology
2214
Store at 4°C. Do not freeze.
IgG2a