Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16/CD32 Antibody (93), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP127946H
Description
CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD16/CD32 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FCGR3A | |
Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors) | |
0.25 mL | |
Primary | |
Murine CD16/32 (CD16/Fc gamma III and CD32/Fc gamma II receptors) | |
Store at 4C in the dark. | |
IgG2a |
Immunohistochemistry (Frozen) | |
93 | |
Immunohistochemistry-Frozen | |
CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3 | |
Rat | |
Protein A or G purified | |
RUO | |
2214 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction