Learn More
CD16/CD32 Monoclonal Antibody (93), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen 25016182
Description
Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils. Applications Reported: The 93 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD16/CD32 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P08101, P08508 | |
Fcgr2b, Fcgr3 | |
Affinity chromatography | |
RUO | |
14130, 14131 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
93 | |
PE-Cyanine7 | |
Fcgr3 | |
AI528646; CD16; CD16a; CD16a antigen; CD16B; CD32; CD32 receptor 2; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIIa; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor (CD16a); fc gamma receptor IIB; Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor RII; Fc gamma RIIB; Fc gamma RIIIa; Fc receptor, IgG, low affinity IIb; Fc receptor, IgG, low affinity III; Fc[g]RII; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD16); fc-gamma RII; Fc-gamma RII-b; Fc-gamma RII-c; Fc-gamma RIII; Fc-gamma RIIIa; Fc-gamma RIII-alpha; Fc-gamma-RII; FcgammaRIIB; fc-gamma-RIIB; Fc-gamma-RIIc; FcgammaRIII; FcgammaRIIIA; Fcgr2; Fcgr2a; Fcgr2b; FCGR2C; Fcgr3; FCGR3A; FCGR3B; FcgRII; FCGRIII; FCR-10; Fcr-2; Fcr-3; fcRII; FcRII-b; Fc | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a λ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.