Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD160 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793915PLATES

 View more versions of this product

Catalog No. NB009822


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD160.
Specifications

Specifications

HRP
Antibody Pairs
93.75 pg/mL
11126
RUO
CD160
Innate Immunity, Signal Transduction
Human
CD160
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 6 ng/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD160.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL rabbit anti-CD160 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-CD160 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 200 ng of recombinant CD160.
  • Storage is content dependent.
    Rabbit
    5 Plates
    93.75-6000 pg/mL
    BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit