Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Mouse anti-Human, PE, Clone: BY55, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 12160942

Catalog No. 50-112-2277


Add to cart

Description

Description

Description: This BY55 monoclonal antibody reacts with human CD160, a member of the immunoglobulin superfamily expressed on natural killer cells, natural killer T cells, activated and/or memory CD4 and CD8 T cells, gamma delta T cells, and intraepithelial cells. CD160 has been reported to bind herpes virus entry mediator (HVEM) in a complex with LIGHT and BTLA to deliver an inhibitory signal to CD4 T cells that abrogates proliferation and cytokine production. Three alternative splice variants of CD160 have also been identified in natural killer cells. Applications Reported: This BY55 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BY55 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
Specifications

Specifications

CD160
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
O95971
NK1, NK28, CD160 antigen, CD160 transmembrane isoform, CD160-delta Ig, natural killer cell receptor BY55, natural killer cell receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BY55
PE
CD160
Liquid
CD160
IgM
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
11126
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.