Learn More
CD161 Mouse anti-Human, Alexa Fluor™ 532, Clone: 14F2F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP214844AF532
Catalog No. NP214844A32
Add to cart
Description
Description
CD161 Monoclonal antibody specifically detects CD161 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CD161
14F2F11
Western Blot
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
KLRB1
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody.
0.1 mL
Primary
3820