Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Mouse anti-Rat, Alexa Fluor 700, Clone: 28764, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9766N100UG
This item is not returnable.
View return policy
Description
CD161 Monoclonal specifically detects CD161 in Rat samples. It is validated for Flow Cytometry.Specifications
CD161 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects rat CD161 in ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
Oct-78 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR | |
Purified splenic NK cells from the LEW rat strain | |
100 μg | |
3820 | |
Rat | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title