Learn More
CD161 Mouse anti-Rat, Alexa Fluor 750, Clone: 28764, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9766S100UG
Catalog No. FAB9766S100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD161
Oct-78
Flow Cytometry 0.25-1 ug/10^6 cells
Supernatant
Mouse
IgG1
100 ug
Primary
3820
Rat
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR
Purified splenic NK cells from the LEW rat strain
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects rat CD161 in ELISAs.