Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161/NK1.1 Mouse anti-Mouse, Clone: PK136, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30753210UG

 View more versions of this product

Catalog No. NB122537


Add to cart

Description

Description

CD161/NK1.1 Monoclonal antibody specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Functional Assay
Specifications

Specifications

CD161/NK1.1
PK136
Unconjugated
10 mM PBS with 0.05% BSA
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
CD Mouse spleen and bone marrow cells enriched for NK-1+ cells
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Mouse
Flow Cytometry, Immunofluorescence, Functional Assay
0.2 mg/ml
Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional
Purified
Mouse
IgG2a Kappa
100 μg
Innate Immunity, Signal Transduction
Primary
17059
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit