Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161a, b, c Rat anti-Mouse, PE, Clone: 16.1, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 566323

Catalog No. BDB566323


Add to cart

Description

Description

The 16.1 monoclonal antibody recognizes the C57BL/6 mouse CD161a, CD161b, and CD161c (CD161a, b, c) cell surface molecules, which are also known as Natural killer cell receptor P1A (NKRP1A), NKRP1D, and NKRP1C, respectively. These cell surface molecules are expressed by natural killer (NK) cells and may play a role in NK cell functions. The 16.1 antibody does not crossreact with CD161f (NKRP1F). This antibody likely binds to CD161 molecules expressed by lymphoid cells from some other mouse strains.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
Specifications

Specifications

CD161a, b, c
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CD161a,b,c, Ly-55a, Ly-55d, Ly-55c, NKRP1A/C/D, Klrb1a, Klrb1d, Klrb1c
Mouse NKRP1A/C/D Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
RUO
Primary
Murine
16.1
PE
Affinity Purified
Rat
Rat PVG, also known as HO, Black Hooded IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit