Learn More
CD162 Mouse anti-Human, APC, Clone: TC2, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15720
Catalog No. 50-113-7076
Add to cart
Description
Description
SELPLG is the high affinity counter-receptor for P-selectin on myeloid cells and stimulated T lymphocytes. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin. The organization of the SELPG gene closely resembles that of CD43 and the human platelet glycoprotein GpIb-alpha both of which have an intron in the 5-prime-noncoding region, a long second exon containing the complete coding region, and TATA-less promoters.
Specifications
Specifications
CD162
Monoclonal
APC
SELPLG
Liquid
SELPLG
Human CD162
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TC2
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Q14242
CD162, CLA, PSGL-1, PSGL1, P-selectin glycoprotein ligand 1, cutaneous lymphocyte-associated associated antigen, SELPG, P-Selectin Glycoprotein Ligand 1
Mouse
IgG1
100 tests
4° C
Primary
6404